Anti-Integrin alpha E Mouse Recombinant Antibody [clone: rITGAE/2063]
Catalog # ANTIA249060-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:A530055J10
- Antigen symbol:Integrin alpha E
- Clonality:Recombinant
- Clone:rITGAE/2063
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P38570
- Antigen synonyms:alpha-M290|aM290|alpha-E1|HML 1 antigen|CD 103|Antigen CD103|HML-1 antigen|CD103 antigen|HML1 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:150 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment of human ITGAE protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rITGAE/2063] antibody to Integrin alpha E for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Integrin alpha E
Clonality: Recombinant
Clone: rITGAE/2063
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human