Anti-GFAP Rabbit Recombinant Antibody [clone: ASTRO/1974R]
Catalog # ANTIA248712-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:ALXDRD
- Antigen symbol:GFAP
- Clonality:Recombinant
- Clone:ASTRO/1974R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig,Rabbit,Chicken,Human,Bovine,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P14136
- Antigen synonyms:GFAP_HUMAN|FLJ45472|Glial fibrillary acidic protein|gfapl|cb345|Intermediate filament protein|etID36982.3|FLJ42474|GFAP
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:~50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human GFAP protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [ASTRO/1974R] antibody to GFAP for flow cytometry and IHC-P with samples derived from human, mouse, rat, bovine, porcine, rabbit and chicken.
- Validated applications: Flow cytometry, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: GFAP
Clonality: Recombinant
Clone: ASTRO/1974R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Porcine, Rabbit, Chicken