Order Entry
Austria
ContactUsLinkComponent
Anti-CREBBP Rabbit Polyclonal Antibody
Anti-CREBBP Rabbit Polyclonal Antibody
Catalog # ANTIA304870-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CREBBP Rabbit Polyclonal Antibody
Catalog # ANTIA304870-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CBP
  • Antigen symbol:
    CREBBP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q92793
  • Antigen synonyms:
    CREB binding protein|RTS|RSTS|CBP_HUMAN|KAT3A / CBP|Crebbp|Cyclic AMP responsive enhancer binding protein|KAT3A|CREB-binding protein
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    265 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide from the mid - protein of Human CBP (aa. 1000 - 1100).
  • Tested applications:
    IHC
  • Purification:
    Peptide affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to CREBBP for WB and IHC with samples derived from human.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:1000, IHC: 1:50

Type: Primary
Antigen: CREBBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human