ContactUsLinkComponent
ROX Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
ROX Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Catalog # AATB67004
Supplier:  AAT BIOQUEST
New Product
ROX Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Catalog # AATB67004
Supplier:  AAT BIOQUEST
New Product
Specifications
  • Frame colour:
    White
  • No. of wells:
    96
  • Profile:
    Low
  • Skirt:
    Semi-skirted
  • Well colour:
    White
  • Description:
    ROX Dye qPCR calibration plate
  • Barcoded:
    Yes
  • Pk:
    1
Specifications
About this item
The ROX dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.