EPA Method 552 Halogenated Acetic Mixture 1000 µg/ml in Methyl tert-butyl ether
EHERA50000758MB
New Product
:
- Pk:1 mL
- Description:EPA method 552 halogenated acetic mixture 1000 µg/ml in methyl tert-butyl ether
- Shipping conditions:–18 °C
- Storage temperature:–18 °C