Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Additives and Supplements for Microbiology
Media supplements
Media supplements
Media supplements
Catalog # HIMEFD025-5X5VL
Supplier:
HIMEDIA
New Product
CAS Number:
Expand image
Media supplements
Catalog # HIMEFD025-5X5VL
Supplier:
HIMEDIA
New Product
CAS Number:
Specifications
Pk:
5 Vial
Description:
Vitamino, sufficient for: 5×500 ml
Specifications
About this item
Product Details
Product Details