Order Entry
Austria
ContactUsLinkComponent
DCI Activator, 0,5 M
DCI Activator, 0,5 M
Catalog # SIALL032100-4X2.5L
Supplier:  Merck
New Product
CAS Number:  
DCI Activator, 0,5 M
Catalog # SIALL032100-4X2.5L
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Pk:
    4
  • Description:
    DCI activator
  • Storage temperature:
    RT

Specifications

About this item

Activator solution, containing powerful activators like 4,5-dicyanoimidazole(DCI), 5-(3,5-bis(trifluoromethyl)phenyl)-1H-tetrazole (Activator 42) or5-(Ethylthio)-1H-tetrazole (ETT) in acetonitrile, are mixed with solutions ofphosphoramidites during the extension step. The activator reacts with theamidite group to form a highly reactive intermediate. The intermediate thenforms an internucleotide bond with the detritylated 5′hydroxyl group of thegrowing oligonucleotide chain.

  • Preparation Note: Dicycanoimidazole/Acetonitrile 6/100 (w/v)