ContactUsLinkComponent
pNPP ELISA Substrate
  STMC01917
 :  
undefined
pNPP ELISA Substrate
  STMC01917
 :  
  • Pk:
    120 mL
  • Primary antibody reactivity:
    Human
    Mouse
  • Target protein:
    1917
Substrate for ELISA antibody pair kits.
pNPP (para-nitrophenyl phosphate) is a chromogenic substrate for alkaline phosphatase (ALP) that is widely used in ELISA assays. pNPP ELISA Substrate is for use with STEMCELL's ELISA antibody pair kits and is supplied in a ready-to-use format in a diethanolamine buffer. The reaction of pNPP ELISA Substrate with ALP results in a yellow, water-soluble product with an absorbance that can be read at 405 nm.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').