Order Entry
Austria
ContactUsLinkComponent
 
Organic Reference Standard, Diazinon-oxon
Catalog # EHERDRE-CA12210200
Supplier:  EHRENSTORFER
New Product
CAS Number:  
Organic Reference Standard, Diazinon-oxon
Catalog # EHERDRE-CA12210200
Supplier:  EHRENSTORFER
New Product
CAS Number:  

Specifications

  • Pk:
    10 mg
  • CAS No.:
    962-58-3
  • Description:
    Organic referrence material diazinon-oxon
  • UN No.:
    2810
  • GHS signal:
    Danger
  • Formula:
    C₁₂H₂₁N₂O₄P
  • Molecular weight:
    288,28

Specifications

About this item

Caution: Laboratory use only.