Express Eingabe
Austria
ContactUsLinkComponent
Transmitter Liquisys, 4-Wire CPM223
Transmitter Liquisys, 4-Wire CPM223
  ENDRCPM223
New Product
Transmitter Liquisys, 4-Wire CPM223
  ENDRCPM223
New Product
  • Bezeichnung:
    One-channel-transmitter, pH/ORP, 4-wire with a two-line display
  • VE:
    1
Compact panel device for all industries.
  • More operational safety
  • Reliable self-monitoring
  • Easy to operate and service
  • Reduced maintenance
  • Fit for every application
Liquisys CPM223 is a standard pH/ORP transmitter for all analog and digital Memosens sensors. It improves your operational safety thanks to continuous plausibility, process and sensor checks. Select from numerous hardware and software modules, such as relays or fieldbus communication, to adapt it exactly to your measuring task. This modularity also allows you to upgrade the transmitter at any time. A simple menu and two-point calibration make configuration and operation fast and easy.
Continuous process check system, customized alarm configuration, proven Memosens interface for reliable sensor connection. Sensor check system for pH glass and reference, calibration plausibility check. Intuitive user interface, hot plug & play with pre-calibrated Memosens sensors, direct access for manual contact control.
Automatic cleaning function (with Chemoclean) triggered by alarm or limit switch. Numerous extensions, such as P(ID) controller, timer, etc., allow for flexible adaption to all processes.
Temperature sensor: Display and current output
Design: pH/ORP transmitter in a panel housing (96x96 mm)
Application: Water, waste water, process
Dimension: 96×96×146 mm (3,74x3,74x5,61") (built in depth)
 : Single channel transmitter Liquisys CPM223.