Order Entry
Austria
ContactUsLinkComponent
Bromocresol Purple (BCP) Agar
Bromocresol Purple (BCP) Agar
Catalog # OXOIPO5118A
New Product
CAS Number:  
Bromocresol Purple (BCP) Agar
Catalog # OXOIPO5118A
New Product
CAS Number:  

Specifications

  • Media Format:
    Powder
  • Pk:
    10
  • Description:
    Bromocresol purple (BCP) agar
  • Storage conditions:
    2...12 °C, dark
  • Volume:
    19,0 g

Specifications

About this item

A medium for the isolation and differentiation of lactose positive and lactose negative enterobacteriaceae.