ContactUsLinkComponent
3M™ Emphaze™ AEX Hybrid Purifier
New Product
3M™ Emphaze™ AEX Hybrid Purifier
New Product
  • Catalog No:
  • MMMEEMP101AEX020R
  • MMMEEMP301AEX020R
  • Autoclavable:
  • 121 °C, 30 min for scale up filter and 121 °C, 40 min for production scale filter
  • 121 °C, 30 min for scale up filter and 121 °C, 40 min for production scale filter
  • Inlet/outlet connection:
  • Luer lock
  • Luer lock
  • Max. operating temperature:
  • 40 °C (104 °F)
  • 40 °C (104 °F)
Frequently Bought Together