Order Entry
Austria
ContactUsLinkComponent
AzoDye-3 azide
AzoDye-3 azide
Catalog # AATB2482
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
AzoDye-3 azide
Catalog # AATB2482
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
Storage Temperature:Freezer
 :651.22 g/mol