ContactUsLinkComponent
Amplitube™ Cap Strips for Well Plates
Amplitube™ Cap Strips for Well Plates
Catalog # SIMPT323-1N
Supplier:  Simport Scientific
New Product
Amplitube™ Cap Strips for Well Plates
Catalog # SIMPT323-1N
Supplier:  Simport Scientific
New Product
Specifications
  • Colour:
    Natural
  • Description:
    Domed 8-strip cap
  • Pk:
    125
Specifications
About this item
Amplitube™ domed cap strips ensure a perfect closure and a positive seal during thermal cycling and prevent evaporation.
  • 8 domed cap strips
  • Made of FDA compliant polypropylene
  • Free of RNase, DNase, Pyrogen, and DNA
  • Leakproof/evaporation tested
Packaging: Packaged in tamperproof resealable bags.