Order Entry
Austria
Orders LinkContactUsLinkComponent
Conductivity Calibration Solution

Conductivity Calibration Solution

Calibration solution, 74 µS/cm, 25 °C, 500 mL
Catalog # ENDRCLY11-A
Supplier:   Endress+Hauser
Conductivity Calibration Solution
Catalog # ENDRCLY11-A
Supplier:  Endress+Hauser
CAS Number:  
Conductivity Calibration Solution

Calibration solution, 74 µS/cm, 25 °C, 500 mL

CAS Number:  
Price
432,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Precision solution referred to SRM of NIST.

With temperature table for qualified calibration of conductivity measuring systems acc. to ISO.

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Description:
    Calibration solution
  • Conductivity:
    74 µS/cm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.