Order Entry
Austria
ContactUsLinkComponent
Klerwipe™ Sporicide Wipes, Low Residue Peroxide Blended with WFI
Klerwipe™ Sporicide Wipes, Low Residue Peroxide Blended with WFI
Catalog # SHIE6600586
New Product
Klerwipe™ Sporicide Wipes, Low Residue Peroxide Blended with WFI
Catalog # SHIE6600586
New Product

Specifications

  • Description:
    Klerwipe sporicidal low residue peroxide pouch wipe, country AT, CZ, DE, HR, HU, SI, SK
  • L×W:
    200×200 mm
  • Seq:
    01
  • Efficacy:
    Wipe_efficacy-21399680|Wipe_efficacy-21399682|Wipe_efficacy-21399679|Wipe_efficacy-21399678
  • Pk:
    200

Specifications

About this item

Sterile 100% polyester wipes impregnated with a very low residue blend of 6% hydrogen peroxide.

  • Ready-to-use
  • Very low residue rotational sporicide wipe
  • For small surfaces

The biocide blend is 0,2 µm filtered and sterile filled under Grade A laminar flow and double bagged in a Grade B cleanroom.

Packaging: Packed in a resealable pouch.