Order Entry
Austria
Orders LinkContactUsLinkComponent
Decoquinate

Decoquinate

100 mg
Catalog # ACSDSB50590.100MG
Supplier:   CPACHEM
Decoquinate
Catalog # ACSDSB50590.100MG
Supplier:  CPACHEM
CAS Number:  18507-89-6
Decoquinate

100 mg

CAS Number:  18507-89-6
Price
270,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
  • CAS Number:18507-89-6
  • Formula:C₂₄H₃₅NO₅
  • MDL Number:MFCD00673686
  • Molecular Weight:417.55 g/mol
  • Melting Pt:242...246 °C
  • Storage Temperature:Refrigerator
  • EINECS:242-389-1
Specifications

Specifications

  • Pk:
    100 mg

Specifications