Order Entry
Austria
ContactUsLinkComponent
Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: MOPC-173] (Biotin)
Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: MOPC-173] (Biotin)
Certificates
undefined
Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: MOPC-173] (Biotin)

Specifications

  • Catalog No:
  • STMC60071BT.1
  • STMC60071BT
  • Antigen name:
  • Igg2a Isotype Control
  • Igg2a Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • MOPC-173
  • MOPC-173
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications

Frequently Bought Together