Order Entry
Austria
ContactUsLinkComponent
Anti-TRA-1-81 Mouse Monoclonal Antibody [clone: TRA-1-81] (Alexa Fluor® 488)
Anti-TRA-1-81 Mouse Monoclonal Antibody [clone: TRA-1-81] (Alexa Fluor® 488)
Certificates
undefined
Anti-TRA-1-81 Mouse Monoclonal Antibody [clone: TRA-1-81] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60065AD
  • STMC60065AD.1
  • Antigen name:
  • TRA-1-81
  • TRA-1-81
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • TRA-1-81
  • TRA-1-81
  • Reactivity:
  • Rhesus monkey, Human, Rat
  • Rhesus monkey, Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5420
  • 5420
  • Isotype:
  • IgM kappa
  • IgM kappa

Specifications

Frequently Bought Together