Mouse monoclonal IgG1 antibody against human, rhesus, chimpanzee CD73, PE-conjugated.
- Mesenchymal Stem
- Progenitor cells
The AD2 antibody reacts with human CD73, a glycosyl phosphatidylinositol (GPI)-anchored glycoprotein and ecto-5’-nucleotidase expressed on the surface of subsets of B and T cells, follicular dendritic cells, mesenchymal stem cells, endothelial cells, and epithelial cells. CD73 catalyzes the hydrolysis of 5’-adenosine monophosphate (AMP) to form the bioactive nucleoside adenosine, and plays a pivotal role in the activation of P1 adenosine receptors by regulating extracellular adenosine concentrations. CD73 also appears to function as a co-signaling molecule on T cells and as an adhesion molecule mediating lymphocyte interactions with the endothelium and follicular dendritic cells. CD73 is used as a marker for lymphocyte differentiation, its expression increasing during development. It is also a useful marker for identifying undifferentiated mesenchymal stem cells. CD73 is highly expressed in many types of human and mouse cancers, and has been implicated in the control of tumor growth.
Type:
Antigen: CD73
Clonality: Monoclonal
Clone: AD2
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee;Human;Macaque;Rhesus monkey
- Catalog No:
- STMC60044PE
- STMC60044PE.1
- Antigen name:
- CD73
- CD73
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- AD2
- AD2
- Reactivity:
- Rhesus monkey, Macaque, Chimpanzee, Human
- Rhesus monkey, Macaque, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 4907
- 4907
- Isotype:
- IgG1 kappa
- IgG1 kappa