Order Entry
Austria
ContactUsLinkComponent
Anti-CD16 Mouse Monoclonal Antibody [clone: 3G8] (Alexa Fluor® 488)
Anti-CD16 Mouse Monoclonal Antibody [clone: 3G8] (Alexa Fluor® 488)
Certificates
undefined
Anti-CD16 Mouse Monoclonal Antibody [clone: 3G8] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60041AD.1
  • STMC60041AD
  • Antigen name:
  • CD16
  • CD16
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • 3G8
  • 3G8
  • Reactivity:
  • Baboon, Capuchin monkey, Cynomolgus monkey, Sooty mangabey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Macaque, Chimpanzee, Human, Squirrel monkey
  • Baboon, Capuchin monkey, Cynomolgus monkey, Sooty mangabey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Macaque, Chimpanzee, Human, Squirrel monkey
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2214
  • 2214
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together