Order Entry
Austria
ContactUsLinkComponent
Anti-TCR Gamma/Delta Hamster Monoclonal Antibody [clone: GL3] (APC)
Anti-TCR Gamma/Delta Hamster Monoclonal Antibody [clone: GL3] (APC)
Certificates
undefined
Anti-TCR Gamma/Delta Hamster Monoclonal Antibody [clone: GL3] (APC)

Specifications

  • Catalog No:
  • STMC60104AZ
  • STMC60104AZ.1
  • Antigen name:
  • TCR Gamma/Delta
  • TCR Gamma/Delta
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • GL3
  • GL3
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Hamster
  • Hamster
  • Gene ID:
  • 110066/110067
  • 110066/110067
  • Isotype:
  • IgG2 kappa
  • IgG2 kappa

Specifications

Frequently Bought Together