Order Entry
Austria
ContactUsLinkComponent
Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (Alexa Fluor® 488)
Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (Alexa Fluor® 488)
Certificates
undefined
Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60004AD
  • STMC60004AD.1
  • Antigen name:
  • CD14
  • CD14
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • M5E2
  • M5E2
  • Reactivity:
  • Dog, Squirrel monkey, Capuchin monkey, Pig, Sheep, Cynomolgus monkey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Macaque, Chimpanzee, Human, Bovine
  • Dog, Squirrel monkey, Capuchin monkey, Pig, Sheep, Cynomolgus monkey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Macaque, Chimpanzee, Human, Bovine
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 929
  • 929
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications

Frequently Bought Together