Order Entry
Austria
ContactUsLinkComponent
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114.15.2] (APC)
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114.15.2] (APC)
Certificates
undefined
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114.15.2] (APC)

Specifications

  • Catalog No:
  • STMC100-0291
  • STMC100-0290
  • Antigen name:
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • M5/114.15.2
  • M5/114.15.2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 14961/14969
  • 14961/14969
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together