Order Entry
Austria
ContactUsLinkComponent
Anti-CD25 Mouse Monoclonal Antibody [clone: BC96] (Alexa Fluor® 488)
Anti-CD25 Mouse Monoclonal Antibody [clone: BC96] (Alexa Fluor® 488)
Certificates
undefined
Anti-CD25 Mouse Monoclonal Antibody [clone: BC96] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60158AD
  • STMC60158AD.1
  • Antigen name:
  • CD25
  • CD25
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • BC96
  • BC96
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Rhesus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3559
  • 3559
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together