Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD45RO Mouse Monoclonal Antibody [clone: UCHL1] (PE)
 
undefined
Anti-CD45RO Mouse Monoclonal Antibody [clone: UCHL1] (PE)

 

  • Catalog No:
  • STMC60097PE
  • STMC60097PE.1
  • Antigen name:
  • CD45RO
  • CD45RO
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • UCHL1
  • UCHL1
  • Reactivity:
  • Common marmoset, Chimpanzee, Human
  • Common marmoset, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5788
  • 5788
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

 

Frequently Bought Together