Order Entry
Austria
ContactUsLinkComponent
Anti-Igg2b Isotype Control Mouse Monoclonal Antibody [clone: MPC-11] (APC)
Anti-Igg2b Isotype Control Mouse Monoclonal Antibody [clone: MPC-11] (APC)
Certificates
undefined
Anti-Igg2b Isotype Control Mouse Monoclonal Antibody [clone: MPC-11] (APC)

Specifications

  • Catalog No:
  • STMC60072AZ
  • STMC60072AZ.1
  • Antigen name:
  • Igg2b Isotype Control
  • Igg2b Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • MPC-11
  • MPC-11
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together