Order Entry
Austria
ContactUsLinkComponent
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-4] (PE)
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-4] (PE)
Certificates
undefined
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-4] (PE)

Specifications

  • Catalog No:
  • STMC60029PE
  • STMC60029PE.1
  • Antigen name:
  • CD4
  • CD4
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • RM4-4
  • RM4-4
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12504
  • 12504
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together