Order Entry
Austria
ContactUsLinkComponent
Anti-Gr-1 (Ly-6G/Ly-6C) Rat Monoclonal Antibody [clone: RB6-8C5] (FITC)
Anti-Gr-1 (Ly-6G/Ly-6C) Rat Monoclonal Antibody [clone: RB6-8C5] (FITC)
Certificates
undefined
Anti-Gr-1 (Ly-6G/Ly-6C) Rat Monoclonal Antibody [clone: RB6-8C5] (FITC)

Specifications

  • Catalog No:
  • STMC60028FI.1
  • STMC60028FI
  • Antigen name:
  • Gr-1 (Ly-6G/Ly-6C)
  • Gr-1 (Ly-6G/Ly-6C)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • RB6-8C5
  • RB6-8C5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 17067/546644
  • 17067/546644
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together