Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD56 (NCAM) Mouse Monoclonal Antibody [clone: HCD56] (PE)
 
undefined
Anti-CD56 (NCAM) Mouse Monoclonal Antibody [clone: HCD56] (PE)

 

  • Catalog No:
  • STMC60021PE.1
  • STMC60021PE
  • Antigen name:
  • CD56 (NCAM)
  • CD56 (NCAM)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • HCD56
  • HCD56
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4684
  • 4684
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together