About this item
Hamster (Armenian) monoclonal IgG antibody against mouse CD11c, Alexa Fluor® 488-conjugated.
- Dendritic cells
The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor. Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cell proliferation, leukocyte recruitment, and phagocytosis. In mice, CD11c is expressed on dendritic cells, macrophages, monocytes, granulocytes, NK cells, and a subset of T cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ mouse CD11c positive selection kit II (Catalog #18780).
Type: Anti-CD11c Hamster Monoclonal Antibody [clone: N418] (Alexa Fluor® 488)
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: Alexa Fluor® 488
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- STMC60002AD
- STMC60002AD.1
- Antigen name:
- CD11c
- CD11c
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 488
- Alexa Fluor® 488
- Clone:
- N418
- N418
- Reactivity:
- Mouse
- Mouse
- Host:
- Hamster
- Hamster
- Gene ID:
- 16411
- 16411
- Isotype:
- IgG
- IgG