Order Entry
Austria
ContactUsLinkComponent
Acetic Acid (31)
Acetic Acid (31)
Catalog # CMPLJPH123
New Product
CAS Number:  
Acetic Acid (31)
Catalog # CMPLJPH123
New Product
CAS Number:  

Specifications

  • Concentration:
    5 mol/L
  • Pk:
    100 mL
  • CAS No.:
    64-19-7
  • Description:
    Acetic acid (31)
  • Shipping conditions:
    Store under normal laboratory conditions, at temperatures between 15...25 °C
  • UN No.:
    2790
  • GHS signal:
    Danger
  • Shelf life:
    24 months

Specifications

About this item

The standard is specially intended for use in testing Japanese pharmacopoeia and monographs. This solution was produced according of Japanese pharmacopoeia specifications using reagent quality components and water meeting the Japanese pharmacopoeia monograph requirement for purified water.

Certifications: ISO 17034, 17025, 9001 certified.