Order Entry
Austria
ContactUsLinkComponent
 
Anti-Igg1 Isotype Control Rat Monoclonal Antibody [clone: RTK2071] (Biotin)
 
undefined
Anti-Igg1 Isotype Control Rat Monoclonal Antibody [clone: RTK2071] (Biotin)

 

  • Catalog No:
  • STMC60075BT
  • STMC60075BT.1
  • Antigen name:
  • Igg1 Isotype Control
  • Igg1 Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • RTK2071
  • RTK2071
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together