- Antigen name:Igm Isotype Control
- Clonality:Monoclonal
- Clone:MM-30
- Conjugation:Alexa Fluor® 488
- Host:Mouse
- Isotype:IgM kappa
- Pk:100 µG
Mouse monoclonal IgM, kappa isotype control antibody, Alexa Fluor® 488conjugated.
The MM-30 antibody (IgM, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of an appropriate isotype control helps confirm the specificity of the antigen-specfic antibody and indicated non-specific binding that may result from binding to Fc receptors or other cell components. The MM-30 antibody reportedly recognises keyhole limpet hemocyanin. This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining).
Type:
Antigen: Igm Isotype Control
Clonality: Monoclonal
Clone: MM-30
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: