Order Entry
Austria
ContactUsLinkComponent
Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60R] (PE)
Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60R] (PE)
Certificates
undefined
Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60R] (PE)

Specifications

  • Catalog No:
  • STMC60064PE.1
  • STMC60064PE
  • Antigen symbol:
  • TRA-1-60
  • TRA-1-60
  • Antigen name:
  • Podocalyxin
  • Podocalyxin
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • TRA-1-60R
  • TRA-1-60R
  • Reactivity:
  • Rabbit, Rhesus monkey, Human
  • Rabbit, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5420
  • 5420
  • Isotype:
  • IgM kappa
  • IgM kappa

Specifications

Frequently Bought Together