Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD11b Rat Monoclonal Antibody [clone: M1/70] (Alexa Fluor® 488)
 
undefined
Anti-CD11b Rat Monoclonal Antibody [clone: M1/70] (Alexa Fluor® 488)

 

  • Catalog No:
  • STMC60001AD
  • STMC60001AD.1
  • Antigen name:
  • CD11b
  • CD11b
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • M1/70
  • M1/70
  • Reactivity:
  • Baboon, Rabbit, Rhesus monkey, Chimpanzee, Human
  • Baboon, Rabbit, Rhesus monkey, Chimpanzee, Human
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16409
  • 16409
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

 

Frequently Bought Together