About this item
Mouse monoclonal IgG1 antibody against human, rhesus, sooty mangabey CD123 (IL-3Rα), biotin-conjugated.
- Dendritic cells
- Granulocytes
- Subsets
- Hematopoietic stem
- Progenitor cells
- Myeloid cells
The 6H6 antibody reacts with human CD123 (IL-3 receptor subunit α), an ~70 kDa type I transmembrane glycoprotein belonging to the type I cytokine receptor family (type 5 subfamily) and the immunoglobulin (Ig) superfamily. CD123 constitutes the ligand-binding α chain of the heterodimeric IL-3 receptor. CD123 binds IL-3 with low affinity per se, but when associated with CD131 (the signal-transducing β chain of the receptor), high-affinity binding of IL-3 is observed. CD123 is expressed by hematopoietic progenitor cells, endothelial cells, basophils, eosinophils, mast cells, monocytes, macrophages, dendritic cells, megakaryocytes, a subset of B cells, and by neutrophils if cultured in the presence of granulocyte-macrophage colony-stimulating factor (GM-CSF). IL-3 binding to CD123 stimulates proliferation, differentiation, and viability of hematopoietic cells. CD123 is highly expressed in malignancies such as acute leukemia. The 6H6 antibody does not inhibit binding of IL-3 to either CD123 or the IL-3 receptor.
Type:
Antigen: CD123 (IL-3Rα)
Clonality: Monoclonal
Clone: 6H6
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rhesus monkey, Sooty mangabey
Specifications
- Catalog No:
- STMC60110BT.1
- STMC60110BT
- Antigen name:
- CD123 (IL-3Ra)
- CD123 (IL-3Ra)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- 6H6
- 6H6
- Reactivity:
- Sooty mangabey, Rhesus monkey, Human
- Sooty mangabey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3563
- 3563
- Isotype:
- IgG1 kappa
- IgG1 kappa