Order Entry
Austria
ContactUsLinkComponent
Anti-SSEA-4 Mouse Monoclonal Antibody [clone: MC-813-70] (PE)
Anti-SSEA-4 Mouse Monoclonal Antibody [clone: MC-813-70] (PE)
Certificates
undefined
Anti-SSEA-4 Mouse Monoclonal Antibody [clone: MC-813-70] (PE)

Specifications

  • Catalog No:
  • STMC60062PE
  • STMC60062PE.1
  • Antigen name:
  • SSEA-4
  • SSEA-4
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • MC-813-70
  • MC-813-70
  • Reactivity:
  • Rabbit, Rhesus monkey, Chicken, Human, Dog, Rat, Mouse, Cat
  • Rabbit, Rhesus monkey, Chicken, Human, Dog, Rat, Mouse, Cat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 330401
  • 330401
  • Isotype:
  • IgG3 kappa
  • IgG3 kappa

Specifications

Frequently Bought Together