Order Entry
Austria
ContactUsLinkComponent
Anti-CD11c Hamster Monoclonal Antibody [clone: N418] (PerCP-Cy5.5)
Zertifikate
undefined
Anti-CD11c Hamster Monoclonal Antibody [clone: N418] (PerCP-Cy5.5)

Spezifikationen

  • Catalog No:
  • STMC60002PS
  • STMC60002PS.1
  • Antigen name:
  • CD11c
  • CD11c
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Clone:
  • N418
  • N418
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Hamster
  • Hamster
  • Gene ID:
  • 16411
  • 16411
  • Isotype:
  • IgG
  • IgG

Spezifikationen

Frequently Bought Together