Order Entry
Austria
ContactUsLinkComponent
 
Anti-ACE2 Mouse Monoclonal Antibody [clone: AC18F] (ATTO 647N)
Catalog # STMC100-0758
undefined
Anti-ACE2 Mouse Monoclonal Antibody [clone: AC18F] (ATTO 647N)
Catalog # STMC100-0758

Specifications

  • Antigen name:
    Angiotensin Converting Enzyme 2
  • Antigen symbol:
    ACE2
  • Clonality:
    Monoclonal
  • Clone:
    AC18F
  • Conjugation:
    ATTO 647N
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Gene ID:
    59272
  • Pk:
    100 Tests

Specifications

About this item

Mouse monoclonal IgG1 antibody against human angiotensin-converting enzyme 2 (ACE2).

  • Airway cells
  • Epithelial cells

The AC18F antibody reacts with angiotensin-converting enzyme 2 (ACE2), a type I integral membrane protein, which includes a short C-terminal cytoplasmic domain made of 22 amino acids, a transmembrane domain, and a large N-terminal ectodomain made of 740 amino acids. The N-terminal domain of ACE2 contains a catalytic zinc-binding motif, which shares a 42% similarity to the catalytic binding domain of ACE. Despite the similarity in catalytic domains, ACE2 functions as a carboxypeptidase, while ACE functions as either a peptidyl dipeptidase or an endopeptidase. In humans, ACE2 is a key regulator of the renin-angiotensin system, where it catalyzes the cleavage of the vasoconstrictor peptide angiotensin II to angiotensin 1-7. Expression of ACE2 can be found on endothelial cells throughout the body, such as in the kidney, heart, and lungs. In the airways leading to the lungs, ACE2 provides the main entry point for SARS-CoV-2 (2019-nCoV). Once the SARS-CoV-2 Spike Protein binds to ACE2, viral fusion with the host cell membrane can occur, resulting in viral RNA genome delivery.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: ACE2
Clonality: Monoclonal
Clone: AC18F
Conjugation: ATTO 647N
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human