Cloud Point Standard, ASTM D2500 and ISO 3015 (Diesel) (PPCPD)
CMPLPPCPD.L25
:
- Pk:250 mL
- CAS No.:68334-30-5
- Description:Cloud point standard
- Shelf life:24 Months
- Storage temperature:15...25 °C
- Density:0,83 g/cm³
- Flash point:62,5 °C
- Test method:ASTM D2500, ISO3015
: ISO 17034, ISO 17025, ISO 9001.
: Store under normal laboratory conditions, at temperatures between 15 to 25 °C.