Heparin-binding epidermal growth factor (EGF)-like growth factor (HBEGF) is a member of the EGF family (Nishi and Klagsbrun). HBEGF promotes blastocyst adhesion to the uterine wall (Iwamoto and Mekada). It also plays a role in smooth muscle cell hyperplasia and brain injury (Nishi and Klagsburn). HBEGF produced by CD4+ T cells promotes wound healing by stimulating migration and proliferation of keratinocytes, fibroblasts, and smooth muscle cells (Blotnick et al.). It binds to EGFR, ErbB4, ErbB2, and ErbB3, activating the PI3K/AKT signaling cascade (Iwamoto and Mekada). HBEGF is produced in a variety of cells, where it contributes to physiological and pathological processes. HBEGF is overexpressed in ovarian, breast, gastric, colorectal, pancreatic, and endometrial cancers, which likely contributes to pathogenesis (Miyata et al.). This product is animal component-free.
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:HBEGF, ACF
- Purity:≥95%