Order Entry
Austria
ContactUsLinkComponent
 
Human Recombinant LIF, ACF
 
undefined
Human Recombinant LIF, ACF

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    LIF, ACF
  • Purity:
    ≥95%

 

Frequently Bought Together