Order Entry
Austria
ContactUsLinkComponent
Human Recombinant IL-1 beta
Human Recombinant IL-1 beta
Certificates
undefined
Human Recombinant IL-1 beta

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IL-1 beta
  • Purity:
    ≥95%

Specifications

Frequently Bought Together