Order Entry
Austria
ContactUsLinkComponent
Human Recombinant bFGF
Human Recombinant bFGF
Certificates
undefined
Human Recombinant bFGF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Basic fibroblast growth factor|FGF-β|FGF2|Fibroblast growth factor-basic|HBGF-2
  • Protein/peptide name:
    bFGF
  • Purity:
    95%

Specifications

Frequently Bought Together