Order Entry
Austria
ContactUsLinkComponent
 
Mouse Recombinant TECK (CCL25)
 
undefined
Mouse Recombinant TECK (CCL25)

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    TECK (CCL25)
  • Purity:
    ≥98%

 

Frequently Bought Together