Human Recombinant TGF-beta 2
Supplier: STEMCELL Technologies
Certificates
About this item
Transforming growth factor (TGF) beta 2 is a member of the TGF-β superfamily and regulates diverse cellular phenotypes. Similar to TGF-β1 and -β3, TGF-β2 signals via serine-threonine kinase type I and II receptors and activates signal transduction via SMAD family proteins, regulating a variety of functions such as cell proliferation, differentiation, wound healing, apoptosis, and metabolism (de Caestecker; Massague; Zuniga et al.). TGF-β2 is important in many developmental processes; for example, mice with TGF-β2 deletions show defects in the development of cardiac, lung, craniofacial, limb, eye, ear, and urogenital systems (Dunker and Kreiglstein).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:TGF-beta 2
- Purity:≥95%
Specifications
Frequently Bought Together