Order Entry
Austria
ContactUsLinkComponent
Human Recombinant Follistatin
Human Recombinant Follistatin
Certificates
undefined
Human Recombinant Follistatin

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    Follistatin
  • Purity:
    ≥95%

Specifications

Frequently Bought Together