Order Entry
Austria
ContactUsLinkComponent
Human Recombinant RANTES (CCL5)
Human Recombinant RANTES (CCL5)
Certificates
undefined
Human Recombinant RANTES (CCL5)

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    RANTES (CCL5)
  • Purity:
    ≥95%

Specifications

Frequently Bought Together