Mouse Recombinant IL-7
About this item
Interleukin 7 (IL-7) is a member of the type I cytokine family that is critical for T and B cell development and survival. It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.). IL-7 binds to a receptor composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways. IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and T regulatory cells (Carrette and Surh), and provides signals to promote survival, proliferation, and differentiation during B cell development (Clark et al.; Corfe and Paige).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
2 Options Available Below
Product Details & Documents
Interleukin 7 (IL-7) is a member of the type I cytokine family that is critical for T and B cell development and survival. It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.). IL-7 binds to a receptor composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways. IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and T regulatory cells (Carrette and Surh), and provides signals to promote survival, proliferation, and differentiation during B cell development (Clark et al.; Corfe and Paige).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- STMC78054.1
- STMC78054
- Protein/peptide name:
- IL-7
- IL-7
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Purity:
- ≥95%
- ≥95%
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.STMC78054.1Protein/peptide nameIL-7Protein/peptide typeRecombinantSpeciesMousePurity≥95%Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.STMC78054Protein/peptide nameIL-7Protein/peptide typeRecombinantSpeciesMousePurity≥95%Marketsource
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



